COD Digestion Vials, Manganese III, Hach
Supplier: Hach
2623415 2623425
CA26234-15PK 912.97 CAD
CA26234-15 CA11016-944
COD Digestion Vials, Manganese III, Hach
Environmental Test Kits
For determination of Chemical Oxygen Demand (COD) by the Manganese III Reactor Digestion Method
Hach Method 10067.
Ordering information: Pack of 25 (pack of 150 vials available). Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accommodate these items.