Nichipet Premium Digital Micropipettors, Nichiryo America
Supplier: Nichiryo America
00-NPP-2 00-NPP-100 00-NPP-10ML 00-NPP-20 00-NPP-10 00-NPP-1000
89128-710EA 513.5 CAD
89128-710 89128-704 89128-708 89128-712 89128-702 89128-706
Nichipet Premium Digital Micropipettors, Nichiryo America
Pipettes Micropipettes
Innovation in durability, strength, stability, and comfort results in the latest model of the Nichipet series.
  • Easy-calibration function provides simple lab calibration
  • Enlarged push button makes it easy to depress
Seal ring for the Premium series is tested dispensing/aspirating 600000 times (2 to 1000 μl). Nozzle top is highly abrasion tolerant (2 to 1000 μl). Non-corrosive ceramic is used for the plunger (100 μl to 10 ml).
A Hyper Blower system, with longer second-push stroke, improves dispensing efficiency (2, 10,and 20 μl). Hand temperatureMore Product Information