Ward's® AP Chemistry Investigation 10: Kinetics: How Long will that Statue Last?
Supplier: ALDON CORP SE
470303-014
470303-014KT 81.39 CAD
470303-014
Ward's® AP Chemistry Investigation 10: Kinetics: How Long will that Statue Last?
Educational Materials Chemistry Educational Materials Applied Chemistry Learning Activities
Meets AP Science Practices 3, 4, 5, 6, and 7
  • Addresses Big Idea 4
  • Materials provided for 30 students working in pairs
  • Investigation includes updated literature aligned with new AP Chemistry curriculum standards
In this lab, chemical kinetics will be investigated as students explore how various conditions involved in a chemical reaction will affect its rate. Students will begin their investigation by studying the effect of the concentration of the acid on the reaction rate and will then consider other variables that may have an effect. By conducting studies in which these variables are altered, students will ultimately be able to determine the effect that each variable has on reaction rates. Includes calcium carbonate (limestone chips), and hydrochloric acid. requires a thermometer and stop watch, available separately.
AP and Advanced Placement Programs are registered trademarks of the College Entrance Examination Board, which was not involved in the production of and does not endorse these products.
Includes:
Limestone Chips
Hydrochloric Acid
Required but Not Provided: Erlenmeyer Flasks, Rubber Stoppers, Volumetric Flasks, DI Water, Syringes, Glass Delivery Tube or rubber tubing, Analytical Balance, Stopwatches, Thermometer, Stir bar or glass stir rod, Gloves, Safety Goggles, Lab Aprons.