pH Buffer Powder Pillows, Hach
Supplier: Hach
1410766 1407995 2227066 2227166 2226964 2226966 2227064 2227195 2226995 2227095 2227164
CA14107-66PK 74.45 CAD
CA14107-66 CA14079-95 CA22270-66 CA22271-66 CA97021-742 CA97021-744 CA97021-746 CA97021-764 CA97021-766 CA97021-770 CA97021-784 CA97021-808
pH Buffer Powder Pillows, Hach
Buffers pH Calibration Buffers
The unit-dose powder pillows assure buffer freshness and eliminate the possibility of contamination
  • Fresh and reliable
  • No danger of contamination
  • Traceable to NIST standards
Each powder pillow prepares 50 mL of buffer solutions of respective pH.