Z-Lys-OBzl benzenesulfonate

Synonyms: Z-Lys-OBzl·benzenesulfonate, Z-Lys-OBzl·benzenesulphonate

C-2210.0005BAEA 305.05 CAD
C-2210.0005BA C-2210.0025BA
Z-Lys-OBzl benzenesulfonate
Z-Lys-OBzl benzenesulfonate

Sequence: Z-Lys-OBzl · benzenesulfonate


For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".

Formula: C₂₇H₃₂N₂O₇S
MW: 528.62 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00077024
CAS Number: 68973-36-4
EINECS: 236-831-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR