Agarose, Ultra Sieve for electrophoresis

Supplier: IBI Scientific

IB70054 IB70055
87004-986EA 290.42 CAD
87004-986 87004-988
Agarose, Ultra Sieve for electrophoresis
Agarose

Ultra Sieve Agarose is designed for high resolution electrophoresis where resolution of polyacrylamide gels are needed. Ultra Sieve Agarose improves clarity of gels and enhances visualization, even at high concentrations. Ultra Sieve Agarose has low viscosity, therefore high concentration gels can be made with high resolving capacity. The high gel strength, low concentration of 1.0-1.5% can also be made. Using TAE buffer, make a 1.8% gel for 400-1200bp, 3.0% gel for 150-800bp, and 4.5% gel for 15-400bp. To achieve the best resolution, store ultra sieve gels at +4-8°C for 30 minutes prior to use.

Melting Pt: 60…90 °C
Storage Temperature: Ambient
MDL Number: MFCD00081294
CAS Number: 9012-36-6
EINECS: 232-731-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=526c20a29610acf6900f898ff245c5602558c785&vid=ae34e4c76a4d7838d4dc9501397f6741b1b97d5e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Moisture Max. 7%
Ash Max. 0.35%
EEO Max. 0.12
Sulfate Max. 0.11%
Gel Strength @1.5% (g/cm^2) Min. 600
Gel Strength @ 1.5% (g/cm^2) Min. 1500
Gelling Temperature @1.5% Max. 35.5°C
Melting Temperature @ 1.5% Max. 80.0°C
Dnase assay None Detected
Rnase assay None Detected
Protease assay None Detected
DNA Extraction assay Pass

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR