Agarose for electrophoresis DNase-, RNase-free

Supplier: IBI Scientific

IB70070 IB70071 IB70072 IB70073 IB70074
97066-028EA 173.04 CAD
97066-028 97066-030 97066-032 97066-034 97066-036
Agarose for electrophoresis DNase-, RNase-free
Agarose

IBI Basic Agarose is a molecular biology grade agarose ideal for use in standard electrophoresis procedures such as rapid separation of DNA and RNA fragments, PCR products, preparation of plasmids and for screening, cloning and blotting techniques.


  • Certified for standard electrophoresis procedures
  • Excellent gel strength and clarity
  • Economical alternative for basic electrophoresis procedures
  • Lot-to-lot consistency
  • DNase- and RNase-Free


IBI Basic Agarose is a neutral polysaccharide extracted from the cell walls of certain Rhodophyceae which are also known as agarophyte seaweeds. The structure of the polysaccharide is a galactan, formed by linking agarobioses 1-3, 1-4. This chemical structure gives agarose the capacity to form gels that are very strong even at low concentrations. These gels have a macroreticular structure with a very open mesh which can be adjusted simply by varying the concentration of agarose.


This is a molecular biology certified agarose product that has been designed and refined for use in every day routine electrophoresis applications. IBI Basic agarose provides a more economical choice for procedures that do not involve resolving small (<1000bp) DNA fragments or fragment that are within 15% to 20% in size of each other.

Melting Pt: 60…90 °C
Storage Temperature: Ambient
MDL Number: MFCD00081294
CAS Number: 9012-36-6
EINECS: 232-731-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=526c20a29610acf6900f898ff245c5602558c785&vid=ae34e4c76a4d7838d4dc9501397f6741b1b97d5e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Ash ≤4.50%
Sulfate ≤0.10%
Clarity @ 1.5% (NTU) ≤4NTU
Gel Strength @ 1.0% (g/cm2) ≥1000
Gel Strength @ 1.5% (g/cm2) ≥2200
Gelling Temperature 36°C ± 1.5°C
Melting Temperature 88°C ± 1.5°C
Dnase Assay None Detected
Rnase Assay None Detected
Protease Assay None Detected
Endonuclease Assay None Detected
Gel Analysis Assay Pass


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR