Addition Funnels, Graduated, Ace Glass Incorporated
Supplier: Ace Glass
7318-12 7318-20 7318-08 7318-16 7268-70 7268-60 7268-64 7268-62 7268-66 9499-02 9499-04 9499-05 9499-06 9499-08 9499-12
89058-544EA 901.62 CAD
89058-544 89058-548 89058-650 89058-674 89058-678 89058-702 89058-706 89058-712 89058-774 89074-998 89075-000 89075-002 89075-004 89075-006 89075-008
Addition Funnels, Graduated, Ace Glass Incorporated
Funnels Dropping Funnels
Cylindrical addition funnel with [ST] joint top and bottom, drip stem, double-scale graduated and calibrated up from the stopcock
The drip stem does not extend beyond the joint, thereby eliminating a possible source of breakage. Stopcock is available with 1:5 PTFE plug or PTFE metering plug
Ordering information: For stoppers (not supplied), order 89062-824 (24/40) or 89062-850 (14/20). For replacement 1:5 PTFE stopcock plugs, order 89062-770 (2-bore) or 89062-774 (4-bore). For replacement metering valves, order 89062-806 (2 mm) or 89062-808 (4 mm). Funnel with glass plug is also available.