StablCal® ULR Verification Kit, Hach
Supplier: Hach
StablCal®
2714600 2714600
CA89231-424EA 607.24 CAD
CA89231-424 89231-424
StablCal® ULR Verification Kit, Hach
Standards Turbidity Standards
StablCal Verification Kit includes: 0.10 NTU, 0.30 NTU, 0.50 NTU, 1.0 NTU standards (100 mL each), sililcone oil and oiling cloth, and instructions.
  • Precise, Premixed Formazin Solutions
  • Based on Formazin Primary Standards
  • Enhanced Stability Saves Time and Labor
  • Confidence in results
For verification only. Not for calibration.