Sodium hydroxide 4.5 N volumetric solution

Supplier: Hach
Danger

204053 204053
CA11016-486EA 103.32 CAD
CA11016-486 104085-708
Sodium hydroxide 4.5 N volumetric solution
Sodium hydroxide

For volatile acids determination by the esterification method.


Ordering information: Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accommodate these items.

Formula: NaOH
MDL Number: MFCD00003548
CAS Number: 1310-73-2
UN: 1824
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR