PCH Series Personal Benchtop Cooler/Heater, Grant Instruments
Supplier: GRANT USA INC.
PCH-1 USA PCH-2 USA PCH-3 USA
97025-548EA 4455.6 CAD
97025-548 97025-550 89402-936
PCH Series Personal Benchtop Cooler/Heater, Grant Instruments
Block Heaters Digital Block Heaters
The PCH series of personal benchtop cooler/heaters are ideal for laboratory applications requiring cooling and heating in microtubes within the temperature setting range of –10 to 100°C (14 to 212°F)
  • Cooling/heating setting range from -10°C to 100°C, with very rapid cool down and heat-up times
  • Stability ±0.1°C
  • Choice of three models: capacity for up to 32 microtubes in a combination of two sizes (PCH-1) or up to 20 microtubes of one size (PCH-2 and PCH-3)
  • Convenient integral reaction timer with audible alarm
The PCH series offer a very effectiveMore Product Information