Phenol solution 30 g/L

Supplier: Hach
Danger

211220
CA2112-20EA 65.79 CAD
CA2112-20
Phenol solution 30 g/L
Phenol

For interference removal in several tests.

Formula: C₆H₅OH
Storage Temperature: Ambient
MDL Number: MFCD00002143
CAS Number: 108-95-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR