Large Capacity Laboratory Incubators, Reach-In, Shel Lab, Sheldon
Supplier: Sheldon Manufacturing
SMI31 SMI39
89409-324EA 30064.54 CAD
89409-324 89409-326
Large Capacity Laboratory Incubators, Reach-In, Shel Lab, Sheldon
Incubators Standard Incubators
SHEL LAB large reach-in incubators provide over 30 cu.ft
chamber capacities while minimizing the amount of floor space used. Both models incorporate our microprocessor controller to achieve precise temperature uniformity. An independent, secondary temperature controller offers the added security of over temperature protection. The chamber floors are reinforced to support roller apparatus or shakers. Both models are supplied with six shelves capable of supporting the weight of benchtop instruments. One access port.
Ordering information: Two-year limited warranty.