Chlorine Dioxide Reagent, Hach
Supplier: Hach
2070242 2070142 2070042
CA11016-976EA 59.25 CAD
CA11016-976 CA11016-978 CA11016-980
Chlorine Dioxide Reagent, Hach
Environmental Test Kits
For determination of chlorine dioxide (ClO₂) by the chlorophenol red method.