Disposable Syringe Filters, MicroLiter
Supplier: DWK Life Sciences (KIMBLE)
F13-1020-1GF F13-1045 F13-2020-1GF F13-2045 F13-2045-1GF F13-4045 F17-2020-1GF F25-1020 F25-1045 F25-2020 F25-2045 F25-2045-1GF F25-4045 F30-2020 F13-2020 F30-2045
97048-592PK 523.14 CAD
97048-592 97048-593 97048-598 97048-600 97048-602 97048-604 97048-610 97048-612 97048-614 97048-616 97048-618 97048-620 97048-624 97048-626 97048-628 97048-630 97048-632 97048-634 97048-636 97048-640 97048-596 97048-638
Disposable Syringe Filters, MicroLiter
Filters Syringe Filters
MicroLiter's syringe filters are manufactured with the most advanced construction methods and design features available today.
  • Housings are solvent resistant
  • Clear markings
  • Traceable
The polypropylene housings are solvent resistant and clearly marked for media and pore size. All packaging is marked with lot numbers for complete traceability. One micron glass fiber prefilters are available to extend filter media life from three to seven times the normal usage.
Packaging: All syringe filters are packaged in packs of 100.