Sample Vials and Caps, MicroLiter, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
13-1000 13-1200 13-2000 13-2200 15-1000-12 15-1000-8 15-2000-12 15-2000-8 18-1000-16 18-2000-16 24-1000-20 24-1000-30 24-1000-40 24-2000-20 24-2000-30 24-2000-40 13-1000-S
97047-630PK 66.31 CAD
97047-630 97047-654 97047-678 97047-688 97047-722 97047-724 97047-744 97047-746 97047-748 97047-800 97047-806 97048-012 97048-014 97048-016 97048-029 97048-030 97048-032 97047-632
Sample Vials and Caps, MicroLiter, DWK Life Sciences
Vials
All vials may appear to be created equal, but quality and performance make the difference when analyzing vital samples.
MicroLiter’s product line of sample vials offer superior storage and handling of larger volume samples. Available in clear and amber formats, with polypropylene or phenolic closures and a variety of septa and liner styles.
MicroLiter vials are cleaned and packaged in a certified Class 10000 clean room according to a designed process, developed to eliminate chromatographic errors associated with vials. Critical physical dimensions are fully tested, as well as chemical cleanliness testing to ensure that trace levels of residues are not present. The result of this process are identical, clean, dependable vials, certified to ensure optimal performance and reproducible results.
Certifications: Certified Clean