Stablcal® Turbidity Standards Calibration Kit, 2100N/N IS Turbidimeter, 500 ml bottles, Hach
Supplier: Hach
Stablcal®
2662100 2662100
CA97026-812EA 1646.2 CAD
97026-812 CA97026-812
Stablcal® Turbidity Standards Calibration Kit, 2100N/N IS Turbidimeter, 500 ml bottles, Hach
Standards Turbidity Standards
Stablcal® Turbidity Standard for all Turbidimeters.
  • Precise, Premixed Formazin Solutions
  • Based on Formazin Primary Standards
  • Enhanced Stability Saves Time and Labor
  • Confidence in results
USEPA accepted for calibration purposes. Stablcal® Stabilized Formazin Turbidity Standards are true Formazin dilutions developed for use in any turbidimeter. Kit includes 500 mL bottles of <0.1, 20, 200, 1000 and 4000 NTU standards.