CyaniVer® 5 Cyanide Reagent Powder Pillows, 25 ml, Hach
Supplier: Hach
1404169
CA14041-69PK
119.43
CAD
CA14041-69
CyaniVer® 5 Cyanide Reagent Powder Pillows, 25 ml, Hach
Environmental Test Kits
For cyanide determination by the Pyridine-Pyrazolone method (Hach method 8027)
Sample size: 25mL.
Ordering information: Pack of 100 Powder Pillows.