Bleaching 3 Reagent Powder Pillows, Hach
Supplier: Hach
1429449 1429449
CA1429449PK 101.2 CAD
89230-964 CA1429449
Bleaching 3 Reagent Powder Pillows, Hach
Environmental Test Kits
For aluminum determination by the aluminon method, Hach Method 8012
Range: up to 0.80 mg/L Al.
Ordering information: Pack of 100 powder pillows.