SP Bel-Art Quaver® and Quaverette® Vibrating Spatulas, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
H36718-0000 H36714-0000 H36715-0000
13197-374EA 129.75 CAD
13197-374 13197-376 57901-106 57901-140
SP Bel-Art Quaver® and Quaverette® Vibrating Spatulas, Bel-Art Products, a part of SP
Spatulas
Make accurate additions of precious materials as needed and reduce transfer losses.
  • Toothed thumb wheel vibrates tip
  • Flat blade with curved end
  • Quaver® spatula features a flat blade with curved end
Rotating the toothed thumb wheel vibrates the spatula tip, giving you control over transfer from spatula to weighing dish.
The Quaverette® spatula permits utilization of even smaller quantities and containers. Its narrow V-shaped blade and 21 cm (8¼") length maneuvers easily into small mouth containers and tight corners.
Both are available with a plain stainless steel blade or with a brown Fluo-Kem® Teflon® FEP coating.
Certifications: The coating complies with FDA regulations governing direct food contact applications.