Bromocresol green-methyl red solution pH-indicator
Supplier: Hach
Synonyms: Methyl Red - Bromocresol green solution
2329249 2329232
CA2329249EA 118.18 CAD
CA2329249 CA23292-32 CA11028-008
Bromocresol green-methyl red solution pH-indicator
Bromocresol green-methyl red
pH indicator for alkalinity determination by titration
APHA formulation.
Ordering information: Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accommodate these items.