Connecting Adapters, Freeze Drying, Ace Glass
Supplier: Ace Glass
6696-74 6696-76 6696-77
89055-796EA 129.79 CAD
89055-796 89055-798 89055-800
Connecting Adapters, Freeze Drying, Ace Glass
Adapters Adapters, Joints with Tube Connectors
These glass adapters are used to connect freeze drying vessels to the rubber valves on Virtis, Labconco, and most other commercially available freeze drying units.
  • Fabricated from borosilicate glass
#25 Ace-Thred joint to a tube (with or without flange). All three adapters can be used with 89059-226 Ace-Thred bushing adapters, which must be purchased separately.