PFA Bottles, Savillex
Supplier: SAVILLEX
150-01-0100 150-01-0250 150-01-0500 150-01-1000 150-01-0050
30617-482EA 459.04 CAD
30617-482 30617-484 30617-486 30617-488 89422-786
PFA Bottles, Savillex
Bottles Plastic Bottles
PFA bottles are made with virgin, high-purity Perfluoroalkoxy (PFA) resin and manufactured by a proprietary stretch blow molding process, adapted for fluoropolymer use by Savillex.
  • Non-Cytotoxic
  • Extractables report available from Savillex
The accessory PFA GL45 Transfer Closure is designed for use with PFA or FEP bottles sized 100 to 2000 ml. The closure features two ¼" tube ports (one top and one side) and includes ¼" ferrule nuts. It can be used to connect bottles to the Savillex Acid Purification System, or for other liquid transfer applications.
PFA bottles include a matching PFA closure, and are manufactured and individually poly-bagged in an ISO Class 7 cleanroom. The molding process produces a bottle without burrs or flashing, and results in a leak-proof seal without the need for secondary sealing plugs or liners. PFA bottles have a lower trace metal background, smoother surface finish, easier pouring, superior sealing properties, and can be repeatedly autoclaved. PFA bottles are excellent for storing high-purity chemicals and other high value materials.
Ordering information: Accessories not included.