Stablcal® Turbidity Standards, 1.0 to 40 NTU, Hach
Supplier: Hach
2659849 2659949 2660149 2746353 2660153 2660142 2659942 2659842 2746356
CA2659849EA 363.91 CAD
CA2659849 CA26599-49 CA26601-49 CA27463-53 CA71002-460 CA11016-416 CA11016-428 CA11016-432 CA11016-440
Stablcal® Turbidity Standards, 1.0 to 40 NTU, Hach
Standards Turbidity Standards
Stablcal® Turbidity Standards for all turbidimeters.
  • Precise, premixed formazin solutions
  • Based on formazin primary standards
  • Enhanced stability saves time and labor
  • Confidence in results
USEPA accepted for calibration purposes. Stablcal® Stabilized Formazin Turbidity Standards are true formazin dilutions developed for use in any turbidimeter