Tubes, Freeze Drying with Screw Cap, Ace Glass
Supplier: Ace Glass
6696-31 6696-32
89055-782EA 65.52 CAD
89055-782 89055-784
Tubes, Freeze Drying with Screw Cap, Ace Glass
Tubes Culture Tubes
These modified culture tubes come complete with a PTFE lined screw-cap and stops on side to prevent 'suck in' under vacuum.
Both sizes fit inside a #25 Ace-thred bushing for use with 89055-766 'umbrella' manifold. Fabricated from borosilicate glass.
Ordering information: Working capacity of tubes is about one half of the capacity listed.