Vessels, Freeze Drying, Heavy Wall, Ace Glass
Supplier: Ace Glass
6696-14 6696-16 6696-12
89055-772EA 172.03 CAD
89055-772 89055-774 89055-776
Vessels, Freeze Drying, Heavy Wall, Ace Glass
Sample Containers General Purpose Containers
These heavy wall vessels are rugged, less likely to implode
Vessels have a 25 mm O.D. neck to fit #25 Ace-Thred on 89055-766 "umbrella" manifold. This vessel has sloped shoulders for easy removal of contents. Fabricated from borosilicate glass.