IntelliCAL™ LDO101 Rugged Luminescent/Optical Dissolved Oxygen (LDO) Probe, Hach
Supplier: Hach
LDO10115 LDO10130
CA11014-086EA 4784.29 CAD
CA11014-086 CA11016-320
IntelliCAL™ LDO101 Rugged Luminescent/Optical Dissolved Oxygen (LDO) Probe, Hach
Electrodes Dissolved Oxygen Electrodes
Hach IntelliCAL™ LDO offers a revolutionary breakthrough in the measurement of dissolved oxygen, eliminating the numerous reliability and maintenance concerns inherent of older DO measurement technologies
IntelliCAL™ LDO101 is a digital, luminescent/optical dissolved oxygen (LDO) probe. The rugged version is available with a 5, 10, 15, or 30 meter cable and is intended for field use. The LDO101 is ideal for measuring dissolved oxygen in wastewater, drinking water and general water quality applications.
IntelliCAL™ Rugged digital probes can reliably test in the harshest environmentalMore Product Information