SP Bel-Art 100-Place Polypropylene Freezer Storage Boxes, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F18851-0011 F18851-0012 F18851-0016
30621-380CS 1496.35 CAD
30621-380 30621-382 30621-384 30621-386 30621-390 30621-392
SP Bel-Art 100-Place Polypropylene Freezer Storage Boxes, Bel-Art Products, a part of SP
Boxes Cryoboxes
100-place polypropylene storage boxes provide compact, safe storage for 0.5, 1.5 and 2.0 mL tubes and cryo vials.
  • Polypropylene construction
  • Available in a variety of color choices
  • Hinged lids with snap tabs
  • Alphanumeric labeling
Great for organization on the bench, in the freezer, or during transport, they can be used at temperatures –80°C to +121°C (112°F to +249°F)
Hinged lids with snap tab open easily and close securely, while clear translucent color always lets you see inside. Opaque black boxes are ideal for light sensitive samples. Each box position is labeled with an alphanumeric grid for easy management of individual samples. ‘No slide’ features molded in bottom and lid provide secure stacking.
Unlike cardboard boxes they will not break down if exposed to moisture or heavy use. Five color choices let you visually sort and organize samples of different types.