Phenol red solution pH-indicator

Supplier: Hach

Synonyms: 3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide

21132 21149
CA97026-640EA 64.89 CAD
CA97026-640 CA11025-040
Phenol red solution pH-indicator
Phenol red

pH Indicator Solution


  • Color range: pH 6.5-8.5

Formula: C₁₉H₁₄O₅S
MW: 354.38 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003552
CAS Number: 143-74-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR