Falcon® Snap Caps for Test Tubes, Corning
Supplier: Corning
Falcon®
352032 352032
CA60819-251CS 519.81 CAD
CA60819-251 60819-251
Falcon® Snap Caps for Test Tubes, Corning
Closures Test/Sample Tube Closures
Falcon® snap caps are designed for use with 12×75 mm test tubes
These polyethylene caps feature a patented, two-position fit, loose for aerobic culturing, tight for sealing. Sterile.
Ordering information: Snap caps are packed 500 per pack, 2000 per case.