Drying Assembly with Aluminum Holder, Ace Glass
Supplier: Ace Glass
6699-02 6699-20 6699-15 6699-10 6699-04
89055-806EA 254.4 CAD
89055-806 89055-810 89055-812 89055-814 89059-364
Drying Assembly with Aluminum Holder, Ace Glass
Desiccators Desiccators, Glass
This drying assembly, originally designed for Iyophilizing and storing frozen slices of tissue for quantitative histochemistry, can also be used as a vacuum tube when combined with aluminum holders by Nelson and Wakefield (see the Journal of Neuropathology and Exper
Neurology, Vol. XXVII, No. 2, April 1968). The joint is a [ST] 40/50. The aluminum holder measures 25 x 90 x 3 mm, with two rows of eight holes, each 8 mm in diameter. The assembly is furnished with two microscope slides. Hose connection is for use with 5/18-inch I.D. tubing. Fabricated from borosilicate glass.