Fume Hood Shelf, PVC or Polyethylene, 3 to 5 Feet, White, TrippNT
Supplier: TrippNT
50215 50315 50216 50217
89408-444EA 292.82 CAD
89408-444 76203-894 76203-896 76203-898 76204-396 76204-398
Fume Hood Shelf, PVC or Polyethylene, 3 to 5 Feet, White, TrippNT
Furniture Shelves
TrippNT's Fume Hood Shelves come in three sizes in either PVC plastic or extra tough high density polyethylene (HDPE) to keep your hood neat and clean and safe.
  • Shelf comes in 3, 4 or 5-foot lengths
  • Designed to hold, contain, and store supplies normally used in the hood or contamination box
  • No assembly required—comes ready out of the box
  • Keep hazardous materials safe, usable and off of lab floor with these specially designed shelves with back cut outs
  • Laminar flow is free to pull and keep hood safe
  • Choose from PVC or HDPE plastic (high density polyethylene)
Special shelves to lift items off hood or bench floor keeping work area clear and uncluttered. Especially useful to allow laminar air flow or use as a step shelf over smaller items. Eliminates clutter hazards by keeping the work area clear and supplies raised off floor with rear baffle open.
Clears clutter from the work area not necessary to the experiment. Keep chemicals, supplies and reagent out of the way so you can work. Easy to clean with light hexanes or soap and water to "like new" condition for years of service.