Air Sampling System, C.A.R.B. Octopus Glass, Ace Glass
Supplier: Ace Glass
7489-04 7489-08 7489-20 7489-22 7489-24 7489-26
89084-284EA 1308.71 CAD
89084-284 89084-286 89084-288 89084-290 89084-292 89084-294
Air Sampling System, C.A.R.B. Octopus Glass, Ace Glass
Samplers Air Samplers
These vertical systems fit more easily into air sampling stations, trailers and cabinets, and provide a more efficient methodology of sample collection
Both the 4-port and 8-port manifolds are available with either a glass sampling cane with an inverted glass cone, or with a straight glass cane with a polyethylene inverse funnel on top. Approximate I.D. of the sampling tube is 25 mm (1 inch). Approximate overall height of four-port system is 150 cm (60 inches). Approximate overall height of eight-port system is 155 cm (62 inches). Fabricated from borosilicate glass. (C.A.R.B. = CaliforniaMore Product Information