Leakbuster™ 3 Specimen Container, Starplex®
Supplier: Starplex Scientific
TW902-O TW902-O-OR TW952-O TW902-O TW902-O-OR
CA11010-421CS 168.63 CAD
97035-688 CA11010-421 10834-132 10787-056 CA97035-688
Leakbuster™ 3 Specimen Container, Starplex®
Sample Containers General Purpose Containers
This new specimen container features a tamper evident cap that ensures proper cap application by providing visual alignment points, as well as audible and distinct tactile closure confirmation
The textured, flat cap provides an additional writing surface in a stackable format. Each container is lot numbered and features clear graduations.
Ordering information: Leakbuster™ 3 is also available in a clear plastic O.R. wrap format and a Midstream Kit format with a castille wipe enclosed in a clear plastic peel pack.