SPADNS Fluoride Reagent Solution, Hach
Supplier: Hach
44453
44449
CA444-53EA
128.63
CAD
CA444-53
CA444-49
SPADNS Fluoride Reagent Solution, Hach
Environmental Test Kits
SPADNS Fluoride Reagent Solution
USEPA accepted for reporting for drinking and wastewater analysis (distillation required) using Hach Method 8029, Fluoride, SPADNS Method Range: 0.02 to 2.00 mg/L.
Ordering information: Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accomodate these items.