Distillation Receivers, Separate Fractions, Ace Glass
Supplier: Ace Glass
6647-10 6647-04 6647-06 9400-03 9400-04 9400-10
89055-730EA 1535.3 CAD
89055-730 89055-742 89055-744 89074-458 89074-460 89074-476
Distillation Receivers, Separate Fractions, Ace Glass
Distillation Components Distillation Receivers
This distillation receiver is for use where separate fractions are desired without disturbing the distillation pressure
Available with [ST] 14/20 or [ST] 24/40 joints. Receiver flasks on [ST] 14/20 size are 5 mL; on [ST] 24/40 size, 50 mL. [ST] 14/20 size has hose connections for use with 3/8-inch or 5/16-inch I.D. tubing; [ST] 24/40 size has hose connections for use 3/8-inch I.D. tubing. Fabricated from borosilicate glass.
Ordering information: Complete [ST] 14/20 size includes 89074-458 vacuum adapter; 89074-460 distributor; four 89074-584 flasks (5 mL); and five 89059-608 DupontMore Product Information