Reflux Apparatus, Continuous, Ace Glass
Supplier: Ace Glass
6620-12 6620-14 6620-40 6620-45
89055-716EA 1486.69 CAD
89055-716 89055-718 89055-720 89055-722
Reflux Apparatus, Continuous, Ace Glass
Distillation Components Distillation Sets
This continuous reflux apparatus is use to keep solvents pure and free of oxygen
The lower drain is centered for complete drainage of head. The included distillation column is 250 mm long, vacuum jacketed, and unsilvered. The included Allihn condenser is 250 mm. The adapter at top has two hose connections (for use with 3/8-inch I.D. tubing) for maintaining an inert atmosphere. The stopcocks are 2 mm bore PTFE. The joint on the lower drain arm is a [ST] 14/20; all others are [ST] 24/40.
The unit, designed by Dr. Philip Fuchs, Purdue University Chemistry Department, West Lafayette, IN., is available in a 1- or 2-liter size. Fabricated from borosilicate glass.
It features a compact, upright design that allows multiple units to be assembled in small areas. The storage head has an overflow high enough to allow storage of solvent for retrieval by syringe through upper part, or through the lower double oblique stopcock.
Ordering information: Complete 1L size includes 89055-716 Storage Head (500 mL); 89060-426 Vacuum Jacketed 250mm Unsilvered Column; 89083-320 Allihn Condenser ([ST]24/40, 250mm); 89056-464 Flask ([ST] 24/40, 1 L); 89076-026 heating mantle (1 L); 89051-412 hose connection adapter ([ST] 24/40); and 89062-824 full-length hollow glass stopper ([ST] 24/40). Complete 2L size includes 89055-718 Storage Head (1 L); 89060-426 Vacuum Jacketed 250mm Unsilvered Column; 89083-320 Allihn Condenser ([ST]24/40, 250mm); 89057-200 Flask ([ST] 24/40, 2 L); 89076-028 heating mantle (2 L); 89051-412 hose connection adapter ([ST] 24/40); and 89062-824 full-length hollow glass stopper ([ST] 24/40). For optional septa (12 pack), order 89064-960. Shorter versions can fabricated; contact your Avantor representative for details.