Extraction Apparatus, for Liquids with Ether, Ace Glass
Supplier: Ace Glass
6840-02 6840-04 6840-06 6840-08 6840-10 6840-15 6840-17 6840-19 6840-21 6840-23 6840-30 6840-35 6840-40 6840-45 6840-50
89056-060EA 284.83 CAD
89056-060 89056-062 89056-064 89056-066 89056-068 89056-070 89056-072 89056-074 89056-076 89056-078 89056-080 89056-082 89056-084 89056-086 89056-088
Extraction Apparatus, for Liquids with Ether, Ace Glass
Extractors Liquid-Liquid Extractors
This apparatus is designed for the extracting of liquids with ether
The dispersion of the ether is accomplished via the use of a fritted disc, 70-100 micron, which is attached to the base of the funnel tube. The apparatus has a [ST] 24/40 on the bottom of the side arm and a [ST] 40/50 joint at the top. Five different capacities are available. The hose barbs on the condenser are for use with 7/16-inch or 3/8-inch I.D. tubing. Fabricated from borosilicate glass.
Ordering information: Complete item consists of flask, condenser, distributor, and extraction chamber. Individual componentsMore Product Information