Pocket Light Meter, Sper Scientific
Supplier: Sper Scientific
840010C
97035-924EA 510.78 CAD
97035-924
Pocket Light Meter, Sper Scientific
Light Meters
Easily transported in a shirt pocket, this tiny light meter is less than ¾” thick, and weighs only 120g (4.2oz.)
The Pocket Light Meter is certified, and includes a N.I.S.T. traceable certificate of calibration. The unit is also supplied with a 5-year warranty.
The controls, display, and sensor are all neatly contained and protected within the folding case with directions printed inside the cover. When open the sensor may be extended for remote use or snaps into a fixed position atop the case. Unit features a bar graph display, auto power off, and hold functions. It also indicates low battery and over range. Powered by 2 button cell batteries (included).