IntelliCAL™ MTC101 Standard Gel Filled ORP Electrodes, Hach
Supplier: Hach
IntelliCAL™
MTC10103 MTC10101 MTC10103 MTC10101
89501-162EA 1273.09 CAD
89501-162 CA89501-162 CA89233-230 89233-230
IntelliCAL™ MTC101 Standard Gel Filled ORP Electrodes, Hach
Electrodes Redox Electrodes
IntelliCAL™ MTC101 is a digital, combination oxidation reduction potential (ORP/Redox) probe with a non-refillable, gel-electrolyte reference and built-in temperature sensor.
  • Provides fast, stable response in a variety of applications
  • Ultimate traceability in measurement history
  • User alerted when recalibration is needed
  • Flat disc sensor for easy cleaning
  • Low maintenance
  • Ag/AgCl reference system
  • Can be moved between meters without the need to recalibrate or reenter measurement settings
The standard version is available with a 1 or 3 meter cable and is intended for laboratory use. The MTC101 is ideal for measuring absolute mV values in wastewater, drinking water and general applications.
Weight: 0.17 lbs. (0.076 kg)