CDC641T 2-pole Platinum Conductivity Cell by Radiometer Analytical, Hach
Supplier: Hach
B15B001 B15B001
CAB15B001EA 2565.48 CAD
CAB15B001 89500-992
CDC641T 2-pole Platinum Conductivity Cell by Radiometer Analytical, Hach
Electrodes Conductivity Sensors
The Radiometer Analytical CDC641T conductivity cell is a 2-pole conductivity cell with platinum sensor
The probe has a 12 mm diameter glass body and integrated temperature sensor. The cell constant is 0.85 cm-1 and the sensor is platinized. The probe has a fixed cable with MAB6 type connector compatible with Radiometer CDM210 and CDM230 Conductivity Meters. The CDC641T is ideal for continuous measurements in various aqueous and non-aqueous media.