Ammonia Ionic Strength Adjustor (ISA), Powder Pillows, Hach
Supplier: Hach
44471-69 4447169
CA44471-69PK 97.22 CAD
CA44471-69 97021-772
Ammonia Ionic Strength Adjustor (ISA), Powder Pillows, Hach
Buffers Ionic Strength Adjustment Buffers
For use in ammonia determinations by electrode methods
Powder form does not dilute samples. Ideal for determining low ammonia levels.
Ordering information: Pack of 100 Powder Pillows.
Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accomodate these items.