Amino Acid Reagent, Hach
Supplier: Hach
193432 193449
CA1934-32EA 61.49 CAD
CA1934-32 CA1934-49
Amino Acid Reagent, Hach
Environmental Test Kits
Reagent for the determination of silica and phosphate.
Methodology: Phosphorus, reactive (Orthophosphate) Amino Acid Reagent Method 8178