PRP-C18 Reversed-Phase HPLC Replacement Guard Cartridges, Hamilton Company

Supplier: Hamilton
32908 79686 79688 79690
89221-438EA 208.17 CAD
89221-438 30618-526 30618-530 30618-534
PRP-C18 Reversed-Phase HPLC Replacement Guard Cartridges, Hamilton Company
Chromatography Columns
Hamilton guard columns protect your HPLC column.

  • Guard columns are modular and cartridge replacement is easy and tool free
  • The design is optimized to reduce dead volume and prevent extra column band broadening

As with all guard columns these should be installed before the analytical column and after the injection valve.

Guard Cartridges for Stainless Steel HPLC Columns
Housing Material: Anodized aluminum
Connecting Tubing Size and material: ¹/₁₆" Ext. Ø, 0.28 mm Int. Ø, 316 stainless steel
Cartridge Materials: Barrel: CTFE; Sleeve: 304 stainless steel;More Product Information
Order Now

SPECIFICATIONS

Environmentally Preferable
Max. pressure 5000 psi
pH range 1 to 14

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR