Distillation Receivers, Sampling, Ace Glass
Supplier: Ace Glass
6635-10 6635-20
89055-724EA 902.59 CAD
89055-724 89055-746
Distillation Receivers, Sampling, Ace Glass
Distillation Components Distillation Receivers
This receiver is specially designed to receiver draw samples during distillation, or to return the condensed vapor to the flask
Fabricated from borosilicate glass.