PRP-C18 Bulk Resin, Hamilton Company
Supplier: HAMILTON COMPANY
79791
79793
30618-536EA
129.12
CAD
30618-536
30618-540
PRP-C18 Bulk Resin, Hamilton Company
Chromatography Sorbents
HPLC Sorbents
Packing material for reversed phase applications.
Packing material type: PS-DVB functionalized with C18