Total Organic Carbon (TOC) Reagent Sets, LR, MR, HR, Hach
Supplier: Hach
2760345 2760445 2815945
CA27603-45EA 1560.01 CAD
CA27603-45 CA97026-422 CA11024-476
Total Organic Carbon (TOC) Reagent Sets, LR, MR, HR, Hach
Environmental Test Kits
Used for determination of Total Organic Carbon (TOC) either by low range, mid range or high range test 'N Tube' method respectively.
  • Affordable
  • Easy to use
  • Accurate
  • Nonhazardous
Hach Method 10173. Range: 15 - 150 mg/L C.
Cuvette test (Ø 16 mm) completely equipped with all reagents, optimised for reliable measuring results and easy handling.
TNT = Test 'N Tube
16 mm TNT products can be evaluated on DR900 only.
Ordering information: Set includes digestion vials and reagents for 25-50 tests (dependent upon the number of reagent blank tests performed). Requires digestion.
Caution: Hazardous Items:
Items with this mark may be considered hazardous under some shipping conditions.
If necessary, we will change your selected shipping method to accomodate these items.