PFA Vials, Savillex
Supplier: SAVILLEX
200-003-20 200-007-20 200-007-30 200-015-20 200-015-30 200-022-20 200-030-20
30621-024EA 47.02 CAD
30621-024 30621-026 30621-028 30621-030 30621-032 30621-034 30621-036
PFA Vials, Savillex
Vials
Used for sample digestion and/or storage.
  • Service temperature range: –200 to 260 °C (–328 to 500 °F)
The flat exterior bottom surface and high service temperature range of these PFA vials make them ideal for use directly on a hot plate. Manufactured from the highest purity PFA resin, these vials are used for sample digestion prior to ultratrace metals analysis. Also ideal for a wide range of other uses. Available with round or conical shaped interior.
Ordering information: Closures are not included, and must be ordered separately. Vial trays include ID cards.