PFA Jars, Savillex
Supplier: SAVILLEX
100-0090-01 100-0240-01 100-1000-01
30617-474EA 157.34 CAD
30617-474 30617-476 30617-480
PFA Jars, Savillex
Jars General Use Jars
Used for the collection, storage, or disposition of a wide variety of solutions or solids
Service temperature range is –200 to 260°C (–328 to 500°F).
The seamless design, radiused interior corners, and wide mouth opening allow for easy transfer and removal of contents. Knurled closures enable a secure grip. A proprietary PFA molding process produces a container with strong yet translucent walls so contents are visible.
Ordering information: Closures are not included, and must be ordered separately.