Distillation Apparatus, Cyanide, with Float Valve, Ace Glass
Supplier: Ace Glass
6557-50 6557-08 6557-14 6557-15 6557-24
89055-568EA 2615.39 CAD
89055-568 89055-612 89055-614 89055-616 89055-618
Distillation Apparatus, Cyanide, with Float Valve, Ace Glass
Distillation Components Distillation Sets
This cyanide distillation apparatus is used in testing for soluble and insoluble cyanides in water
Two #11 bushings, back to back, hold the threaded glass adapter to the still head by compressing PTFE ferrules against the adjustable depth glass dispersion tube. The frit on the dispersion tube is 10-20 micron. The trap has a top [ST] 19/22 joint. The threaded glass adapter has a 5/16" hose connection. Fabricated from borosilicate glass.
Flask is 1000mL with [ST] 24/40 joint and #7 Ace-Thred that permits variable depth positioning of the fill tube. The fill tube has been designed with a float valve for added protection against “boil back,” or splattering, that occurs with conventional tubes. The still head is jacketed and lengthened for more efficient condensing. Both the head and the cold finger have [ST] 29/42 joints, and there is a #11 Ace-Thred joint on the side arm of the head.
Ordering information: Complete apparatus includes 89056-568 flask; 89055-612 fill tube; 89055-614 distilling head; 89055-616 condenser/finger; 89059-214 #11 Ace-Thred bushing; 89051-494 #11 Ace-Thred glass adapter; 89055-618 dispersion tube with frit; 89055-606 trap; and 89055-610 ferrules pack that includes one #7 and two #11. Individual components may also be ordered separately.